Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OSTF1
Entrez Gene ID
427258
Gene Name
osteoclast stimulating factor 1
Gene Aliases
-
Chromosome Location
Chr.Z: 36854984 - 36875056 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031426.2 NP_001026597.2 8 - 9 613 104
GenBank mRNA AJ720435.1 - 8 - 9 613 104

View Full Product Details