Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
P3H2
Entrez Gene ID
414143
Gene Name
prolyl 3-hydroxylase 2
Gene Aliases
LEPREL1
Chromosome Location
Chr.9: 14138902 - 14200554 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001530.1 NP_001001530.1 9 - 10 1412 65
XM_015291384.2 XP_015146870.1 9 - 10 1496 65
XM_015291385.2 XP_015146871.1 9 - 10 1103 65
GenBank mRNA AY463529.1 - 9 - 10 1412 65
CR733175.1 - 7 - 8 658 65

View Full Product Details