Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYB5A
Entrez Gene ID
414798
Gene Name
cytochrome b5 type A
Gene Aliases
CYB5
Chromosome Location
Chr.2: 91931575 - 91942888 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001748.2 NP_001001748.1 4 - 5 380 78
GenBank mRNA M18539.1 - 4 - 5 357 78
M32293.1 - 4 - 5 381 78

View Full Product Details