Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP3A5
Entrez Gene ID
414832
Gene Name
cytochrome P450 family 3 subfamily A member 5
Gene Aliases
CYP3A37, CYP3A7
Chromosome Location
Chr.14: 4323045 - 4332386 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001751.2 NP_001001751.1 12 - 13 1475 62
GenBank mRNA AJ250337.1 - 12 - 13 1476 62

View Full Product Details