Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCNB1
Entrez Gene ID
415400
Gene Name
cyclin B1
Gene Aliases
CCNB2, cycB2
Chromosome Location
Chr.10: 7264606 - 7272854 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004369.1 NP_001004369.1 3 - 4 328 102
XM_015278892.2 XP_015134378.1 4 - 5 469 102
XM_015278893.2 XP_015134379.1 3 - 4 555 102
GenBank mRNA X62531.1 - - 328 102

View Full Product Details