Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AQP4
Entrez Gene ID
421088
Gene Name
aquaporin 4
Gene Aliases
aquaporin-4
Chromosome Location
Chr.2: 104434445 - 104447281 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004765.2 NP_001004765.2 3 - 4 659 93
NM_001317827.1 NP_001304756.1 3 - 4 659 93
NM_001364409.1 NP_001351338.1 3 - 4 659 93
GenBank mRNA AB190358.1 - 3 - 4 660 93
BX929627.2 - 3 - 4 635 93
BX935883.1 - 2 - 3 291 93
CN383036.1 - 2 - 3 279 93

View Full Product Details