Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POU2AF1
Entrez Gene ID
373994
Gene Name
POU class 2 homeobox associating factor 1
Gene Aliases
CHOBF-1, OBF-1
Chromosome Location
Chr.24: 4343408 - 4355899 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204175.2 NP_989506.1 4 - 5 562 105
XM_015297931.2 XP_015153417.1 4 - 5 972 105
GenBank mRNA AB052869.1 - 4 - 5 520 105
AJ720333.1 - 4 - 5 562 105

View Full Product Details