Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THYN1
Entrez Gene ID
374161
Gene Name
thymocyte nuclear protein 1
Gene Aliases
THY28
Chromosome Location
Chr.24: 2585500 - 2590603 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204287.2 NP_989618.1 2 - 3 182 77
XM_015297937.2 XP_015153423.1 2 - 3 193 77
GenBank mRNA BX950750.2 - 2 - 3 187 77
U34350.1 - 2 - 3 182 77

View Full Product Details