Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL10RB
Entrez Gene ID
395663
Gene Name
interleukin 10 receptor subunit beta
Gene Aliases
IL10R2
Chromosome Location
Chr.1: 106593977 - 106604819 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204857.1 NP_990188.1 2 - 3 167 94
XM_015299252.2 XP_015154738.1 2 - 3 245 94
GenBank mRNA AF082666.1 - 2 - 3 167 94
AM049249.1 - 2 - 3 362 94
AM049250.1 - 2 - 3 362 94

View Full Product Details