Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL10RB
Entrez Gene ID
395663
Gene Name
interleukin 10 receptor subunit beta
Gene Aliases
IL10R2
Chromosome Location
Chr.1: 106593977 - 106604819 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204857.1 NP_990188.1 4 - 5 512 86
XM_015299252.2 XP_015154738.1 4 - 5 590 86
GenBank mRNA AF082666.1 - 4 - 5 512 86
AM049249.1 - 4 - 5 707 86

View Full Product Details