Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIX6
Entrez Gene ID
395843
Gene Name
SIX homeobox 6
Gene Aliases
OPTX2, Six9
Chromosome Location
Chr.5: 54616455 - 54619257 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204994.1 NP_990325.1 2 - 3 1019 118
GenBank mRNA AF050131.1 - 2 - 3 1019 118

View Full Product Details