Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFNB1
Entrez Gene ID
395896
Gene Name
ephrin B1
Gene Aliases
-
Chromosome Location
Chr.4: 1022718 - 1070879 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205035.2 NP_990366.1 3 - 4 744 66
XM_015278194.2 XP_015133680.1 3 - 4 596 66
XM_015278195.2 XP_015133681.1 3 - 4 416 66
GenBank mRNA U72394.1 - 3 - 4 747 66

View Full Product Details