Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GABRA6
Entrez Gene ID
395931
Gene Name
gamma-aminobutyric acid type A receptor alpha6 subunit
Gene Aliases
-
Chromosome Location
Chr.13: 8003525 - 8025111 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205058.1 NP_990389.1 7 - 8 821 115
GenBank mRNA X94343.1 - - 821 115

View Full Product Details