Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSHB
Entrez Gene ID
395937
Gene Name
thyroid stimulating hormone beta
Gene Aliases
TSH-B
Chromosome Location
Chr.26: 3966715 - 3987181 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205063.2 NP_990394.1 - 177 81
XM_015298896.2 XP_015154382.1 - 713 81
XM_025143670.1 XP_024999438.1 - 2161 81
GenBank mRNA AF033495.1 - - 177 81

View Full Product Details