Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ENO1
Entrez Gene ID
396017
Gene Name
enolase 1, (alpha)
Gene Aliases
-
Chromosome Location
Chr.21: 3220608 - 3233067 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205120.1 NP_990451.1 6 - 7 474 62
XM_015296833.2 XP_015152319.2 6 - 7 539 62
XM_015296834.2 XP_015152320.2 6 - 7 470 62
XM_025142487.1 XP_024998255.1 6 - 7 477 62
GenBank mRNA AJ720730.1 - 6 - 7 469 62
D37900.1 - 6 - 7 474 62

View Full Product Details