Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GABRE
Entrez Gene ID
396173
Gene Name
gamma-aminobutyric acid type A receptor subunit epsilon
Gene Aliases
GABRG4
Chromosome Location
Chr.4: 10827137 - 10858894 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205245.1 NP_990576.1 7 - 8 959 82
GenBank mRNA X73533.1 - - 959 82

View Full Product Details