Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGF3
Entrez Gene ID
396267
Gene Name
fibroblast growth factor 3
Gene Aliases
INT-2
Chromosome Location
Chr.5: 17872481 - 17877619 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205327.1 NP_990658.1 1 - 2 221 64
GenBank mRNA Z47555.1 - - 221 64

View Full Product Details