Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HMOX1
Entrez Gene ID
396287
Gene Name
heme oxygenase 1
Gene Aliases
HO-1
Chromosome Location
Chr.1: 52103885 - 52109345 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205344.1 NP_990675.1 1 - 2 35 73
GenBank mRNA HM237181.1 - 1 - 2 123 73
X56201.1 - - 35 73

View Full Product Details