Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LACTB
Entrez Gene ID
415373
Gene Name
lactamase beta
Gene Aliases
-
Chromosome Location
Chr.10: 4602215 - 4609086 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030546.1 NP_001025717.1 4 - 5 849 102
GenBank mRNA AJ720272.1 - 4 - 5 849 102

View Full Product Details