Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
N4BP1
Entrez Gene ID
415739
Gene Name
NEDD4 binding protein 1
Gene Aliases
-
Chromosome Location
Chr.11: 7288050 - 7304747 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030570.1 NP_001025741.1 2 - 3 2118 145
GenBank mRNA AJ719785.1 - 2 - 3 2118 145
CR386283.1 - 1 - 2 451 145

View Full Product Details