Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HNF4beta
Entrez Gene ID
415820
Gene Name
hepatic nuclear factor 4beta
Gene Aliases
-
Chromosome Location
Chr.11: 16993654 - 17014303 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030576.4 NP_001025747.3 2 - 3 373 73
GenBank mRNA AY700582.1 - 2 - 3 477 73
CD739754.1 - 2 - 3 382 73

View Full Product Details