Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HNF4beta
Entrez Gene ID
415820
Gene Name
hepatic nuclear factor 4beta
Gene Aliases
-
Chromosome Location
Chr.11: 16993654 - 17014303 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030576.4 NP_001025747.3 4 - 5 567 87
GenBank mRNA AY700582.1 - 4 - 5 671 87
BU300422.1 - 2 - 3 195 87

View Full Product Details