Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYB5B
Entrez Gene ID
415865
Gene Name
cytochrome b5 type B
Gene Aliases
CYB5-M
Chromosome Location
Chr.11: 18950606 - 18962493 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030581.1 NP_001025752.1 3 - 4 435 103
GenBank mRNA AJ720899.1 - 3 - 4 435 103

View Full Product Details