Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SHISA5
Entrez Gene ID
415948
Gene Name
shisa family member 5
Gene Aliases
-
Chromosome Location
Chr.12: 3651741 - 3662916 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030591.2 NP_001025762.1 2 - 3 135 84
XM_015293043.2 XP_015148529.1 2 - 3 2920 84
GenBank mRNA AJ720705.1 - 2 - 3 136 84

View Full Product Details