Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COPG1
Entrez Gene ID
416014
Gene Name
coatomer protein complex subunit gamma 1
Gene Aliases
COPG
Chromosome Location
Chr.12: 9501312 - 9520387 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030597.2 NP_001025768.1 8 - 9 691 66
GenBank mRNA AJ851498.1 - 8 - 9 692 66

View Full Product Details