Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RHO
Entrez Gene ID
751791
Gene Name
rhodopsin
Gene Aliases
CSNBAD1, OPN2, RDP1, RP4, opsin-2
Chromosome Location
Chr.12: 19916702 - 19919673 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030606.1 NP_001025777.1 2 - 3 528 96

View Full Product Details