Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THG1L
Entrez Gene ID
416242
Gene Name
tRNA-histidine guanylyltransferase 1 like
Gene Aliases
-
Chromosome Location
Chr.13: 12010505 - 12015815 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030616.1 NP_001025787.1 4 - 5 615 146
XM_015293767.1 XP_015149253.1 4 - 5 566 146
XM_015293768.1 XP_015149254.1 3 - 4 231 146
GenBank mRNA AJ720720.1 - 4 - 5 615 146

View Full Product Details