Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMS
Entrez Gene ID
418602
Gene Name
spermine synthase
Gene Aliases
-
Chromosome Location
Chr.1: 120425332 - 120468447 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030803.1 NP_001025974.1 10 - 11 1105 69
GenBank mRNA AJ851504.1 - 10 - 11 1105 69

View Full Product Details