Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDFIP2
Entrez Gene ID
418803
Gene Name
Nedd4 family interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.1: 154396098 - 154442843 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030820.1 NP_001025991.1 4 - 5 441 116
XM_015275343.2 XP_015130829.1 6 - 7 701 116
XM_015275348.2 XP_015130834.1 5 - 6 577 116
XM_015275351.2 XP_015130837.1 5 - 6 641 116
GenBank mRNA AJ851545.1 - 4 - 5 441 116
CR352953.1 - 5 - 6 344 116

View Full Product Details