Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAF3
Entrez Gene ID
419107
Gene Name
TATA-box binding protein associated factor 3
Gene Aliases
-
Chromosome Location
Chr.1: 4825154 - 4938986 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030841.2 NP_001026012.1 5 - 6 2670 79
XM_015293603.2 XP_015149089.1 4 - 5 9760 79
GenBank mRNA AJ851411.1 - 5 - 6 2673 79

View Full Product Details