Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAF8
Entrez Gene ID
419929
Gene Name
TATA-box binding protein associated factor 8
Gene Aliases
TBN
Chromosome Location
Chr.26: 5068715 - 5076380 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030923.1 NP_001026094.1 - 695 92
XM_015298954.2 XP_015154440.1 - 658 92
XM_015298955.2 XP_015154441.1 - 1054 92
XM_025143708.1 XP_024999476.1 - 845 92
GenBank mRNA AJ719313.1 - - 695 92

View Full Product Details