Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
E2F5
Entrez Gene ID
420206
Gene Name
E2F transcription factor 5
Gene Aliases
-
Chromosome Location
Chr.2: 122608814 - 122621360 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030942.1 NP_001026113.1 3 - 4 521 118
XM_015282807.2 XP_015138293.2 3 - 4 423 118
GenBank mRNA AJ720953.1 - 3 - 4 521 118

View Full Product Details