Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRELID3A
Entrez Gene ID
421035
Gene Name
PRELI domain containing 3A
Gene Aliases
SLMO1
Chromosome Location
Chr.2: 97199682 - 97210066 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031011.1 NP_001026182.1 5 - 6 570 117
GenBank mRNA AJ721065.1 - 5 - 6 570 117

View Full Product Details