Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAPRE2
Entrez Gene ID
421105
Gene Name
microtubule associated protein RP/EB family member 2
Gene Aliases
-
Chromosome Location
Chr.2: 107562389 - 107654659 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031017.1 NP_001026188.1 4 - 5 682 137
XM_015282587.2 XP_015138073.2 5 - 6 137
XM_015282589.2 XP_015138075.2 5 - 6 137
XM_015282590.2 XP_015138076.2 5 - 6 137
XM_025147305.1 XP_025003073.1 4 - 5 137
GenBank mRNA AJ720083.1 - 4 - 5 682 137

View Full Product Details