Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H11B
Entrez Gene ID
421172
Gene Name
zinc finger CCCH-type containing 11B pseudogene
Gene Aliases
ZC3H11A
Chromosome Location
Chr.26: 481428 - 496368 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031030.1 NP_001026201.1 - 807 92
XM_015298713.2 XP_015154199.1 - 2391 92
GenBank mRNA AJ720443.1 - - 807 92

View Full Product Details