Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASXL2
Entrez Gene ID
421993
Gene Name
additional sex combs like 2, transcriptional regulator
Gene Aliases
-
Chromosome Location
Chr.3: 104744834 - 104834408 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031096.1 NP_001026267.1 6 - 7 812 70
XM_015285098.2 XP_015140584.1 6 - 7 820 70
XM_015285099.2 XP_015140585.1 8 - 9 711 70
XM_015285100.2 XP_015140586.1 3 - 4 405 70
XM_025148790.1 XP_025004558.1 6 - 7 658 70
GenBank mRNA AJ719496.1 - 6 - 7 812 70

View Full Product Details