Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STK26
Entrez Gene ID
422231
Gene Name
serine/threonine protein kinase 26
Gene Aliases
MST4
Chromosome Location
Chr.4: 3425524 - 3453523 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031117.1 NP_001026288.1 4 - 5 482 72
XM_015278244.2 XP_015133730.1 5 - 6 805 72
GenBank mRNA AJ719477.1 - 4 - 5 482 72

View Full Product Details