Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
UPRT
Entrez Gene ID
422327
Gene Name
uracil phosphoribosyltransferase homolog
Gene Aliases
-
Chromosome Location
Chr.4: 12557477 - 12569504 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031124.1 NP_001026295.1 1 - 2 361 66
GenBank mRNA AJ720786.1 - 1 - 2 361 66

View Full Product Details