Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPATA4
Entrez Gene ID
422571
Gene Name
spermatogenesis associated 4
Gene Aliases
-
Chromosome Location
Chr.4: 44348241 - 44355264 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031138.2 NP_001026309.1 5 - 6 685 147
GenBank mRNA AY761140.1 - 5 - 6 686 147

View Full Product Details