Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NELFA
Entrez Gene ID
422896
Gene Name
negative elongation factor complex member A
Gene Aliases
WHSC2
Chromosome Location
Chr.4: 83173167 - 83196462 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031167.1 NP_001026338.1 5 - 6 1203 114
XM_025149713.1 XP_025005481.1 6 - 7 1033 114
XM_025149714.1 XP_025005482.1 5 - 6 943 114
XM_025149715.1 XP_025005483.1 4 - 5 781 114
GenBank mRNA AJ851513.1 - 5 - 6 1203 114

View Full Product Details