Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
MAP1LC3B
Entrez Gene ID
427559
Gene Name
microtubule associated protein 1 light chain 3 beta
Gene Aliases
MAP1LC3B2
Chromosome Location
Chr.11: 17796381 - 17805114 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031461.1 NP_001026632.1 1 - 2 119 66
GenBank mRNA AJ719776.1 - 1 - 2 119 66

View Full Product Details