Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUDS3
Entrez Gene ID
427713
Gene Name
SDS3 homolog, SIN3A corepressor complex component
Gene Aliases
-
Chromosome Location
Chr.15: 10079884 - 10105673 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031471.1 NP_001026642.1 8 - 9 649 94
XM_015275338.2 XP_015130824.1 8 - 9 3750 94
GenBank mRNA AJ720986.1 - 8 - 9 649 94

View Full Product Details