Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NONO
Entrez Gene ID
428701
Gene Name
non-POU domain containing, octamer-binding
Gene Aliases
-
Chromosome Location
Chr.4: 2268811 - 2283610 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031532.1 NP_001026703.1 6 - 7 1015 71
XM_015278310.2 XP_015133796.1 6 - 7 1137 71
XM_015278311.2 XP_015133797.1 6 - 7 1109 71
XM_015278312.2 XP_015133798.1 5 - 6 1001 71
XM_015278313.2 XP_015133799.1 6 - 7 1021 71
XM_025149746.1 XP_025005514.1 6 - 7 1022 71
GenBank mRNA AJ720639.1 - 6 - 7 1015 71

View Full Product Details