Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APIP
Entrez Gene ID
428860
Gene Name
APAF1 interacting protein
Gene Aliases
-
Chromosome Location
Chr.5: 19085945 - 19098886 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031547.1 NP_001026718.1 7 - 8 774 76
GenBank mRNA AJ719692.1 - 7 - 8 774 76

View Full Product Details