Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MFSD14A
Entrez Gene ID
429081
Gene Name
major facilitator superfamily domain containing 14A
Gene Aliases
HIAT1, MFSD14B
Chromosome Location
Chr.8: 12349287 - 12364548 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031562.1 NP_001026733.1 3 - 4 382 121
GenBank mRNA AJ851716.1 - 3 - 4 382 121

View Full Product Details