Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMF1
Entrez Gene ID
430115
Gene Name
proteasome inhibitor subunit 1
Gene Aliases
-
Chromosome Location
Chr.20: 9867426 - 9873205 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031582.1 NP_001026753.1 5 - 6 625 81
GenBank mRNA AJ720421.1 - 5 - 6 625 81

View Full Product Details