Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
PABPC1
Entrez Gene ID
430997
Gene Name
poly(A) binding protein cytoplasmic 1
Gene Aliases
-
Chromosome Location
Chr.2: 128496930 - 128511621 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031597.1 NP_001026768.1 14 - 15 2391 111
XM_025147342.1 XP_025003110.1 14 - 15 2458 111
GenBank mRNA AJ719881.1 - 14 - 15 2391 111

View Full Product Details