Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LHX3
Entrez Gene ID
373940
Gene Name
LIM homeobox 3
Gene Aliases
lim-3
Chromosome Location
Chr.17: 8343834 - 8350636 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030335.1 NP_001025506.1 3 - 4 447 99
GenBank mRNA AF461699.1 - 3 - 4 477 99
L35570.1 - 3 - 4 447 99

View Full Product Details