Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM27.1
Entrez Gene ID
417042
Gene Name
tripartite motif containing 27.1
Gene Aliases
TRIM27
Chromosome Location
Chr.16: 2514606 - 2518930 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030671.2 NP_001025842.2 2 - 3 494 73
XM_015294957.2 XP_015150443.1 2 - 3 868 73

View Full Product Details