Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NACC2
Entrez Gene ID
417128
Gene Name
NACC family member 2
Gene Aliases
BTBD14A
Chromosome Location
Chr.17: 8388397 - 8438530 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030681.1 NP_001025852.1 3 - 4 1272 111
GenBank mRNA AJ720914.1 - 3 - 4 1272 111

View Full Product Details